Gene Transcripts
- Gene Symbol
- Ppp1cc
- Entrez Gene ID
- 19047
- Gene Name
- protein phosphatase 1, catalytic subunit, gamma isoform
- Gene Aliases
- PP-1G, PP1, dis2m1
- Chromosome Location
- Chr. 5: 122158255 - 122175273 on Build GRCm38
- UniGene ID
- Mm.280784
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_013636.3 |
NP_038664.2
|
6, 7 |
1171 |
|
GenBank mRNA |
AK076009.1 |
- |
6, 7 |
1108 |
|
AK088229.1 |
- |
6, 7 |
1115 |
|
AK147131.1 |
- |
6, 7 |
1173 |
|
AK152422.1 |
- |
6, 7 |
1134 |
|
AK152625.1 |
- |
6, 7 |
1110 |
|
AK153363.1 |
- |
6, 7 |
984 |
|
AK169336.1 |
- |
6, 7 |
1131 |
|
BC008157.1 |
- |
5, 6 |
1279 |
|
BC010613.1 |
- |
6, 7 |
944 |
3589168 |
BC021646.1 |
- |
6, 7 |
1172 |
3487479 |
BC039972.1 |
- |
5, 6 |
1294 |
|
BC085496.1 |
- |
6, 7 |
1110 |
6829867 |
D85137.1 |
- |
6, 7 |
996 |
|
M27071.1 |
- |
6, 7 |
1092 |
|
U53456.1 |
- |
6, 7 |
1088 |
|
- Gene Symbol
- Gm5601
- Entrez Gene ID
- 434233
- Gene Name
- predicted pseudogene 5601
- Gene Aliases
- EG434233
- Chromosome Location
- Chr. 7: 119739888 - 119742244 on Build GRCm38
- UniGene ID
- Mm.280784
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK182169.1 |
- |
1 |
243 |
|
View Full Product Details