Gene Transcripts
- Gene Symbol
- Hspa1b
- Entrez Gene ID
- 294254
- Gene Name
- heat shock 70kD protein 1B (mapped)
- Gene Aliases
- Hsp70-2, Hsp72, Hspa2
- Chromosome Location
- Chr. 20: 4875834 - 4881751 on Build Rnor_6.0
- UniGene ID
- Rn.1950
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_212504.1 |
NP_997669.1
|
1 |
4141 |
|
- Gene Symbol
- Hspa1a
- Entrez Gene ID
- 24472
- Gene Name
- heat shock 70kD protein 1A
- Gene Aliases
- HSP72, Hsp70-1, Hspa1, Hspa1b
- Chromosome Location
- Chr. 20: 2699707 - 2701980 on Build Rnor_6.0
- UniGene ID
- Rn.1950
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
L16764.1 |
- |
|
2168 |
|
View Full Product Details