Gene Transcripts

Gene Symbol
SIAH1
Entrez Gene ID
6477
Gene Name
siah E3 ubiquitin protein ligase 1
Gene Aliases
SIAH1A
Chromosome Location
Chr. 16: 48360531 - 48448440 on Build GRCh38
UniGene ID
Hs.706828
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001006610.1 NP_001006611.1 2 646
NM_003031.3 NP_003022.3 2 274
XM_006721246.1 XP_006721309.1 3 450
XM_011523279.1 XP_011521581.1 6 1790
GenBank mRNA AB451253.1 - 1 157
AB451377.1 - 1 157
AK094663.1 - 1 1611
AK225895.1 - 2 307
AK313861.1 - 2 660
BC035562.1 - 2 274 5224755
BC042550.1 - 2 616 5198176
BC044920.1 - 6 1753 5295925
BX357983.2 - 2 675
BX647064.1 - 1 1218
EF026094.1 - 1 157
U63295.1 - 1 157
U76247.1 - 2 343

View Full Product Details