Gene Transcripts
- Gene Symbol
- Lgals5
- Entrez Gene ID
- 25475
- Gene Name
- lectin, galactose binding, soluble 5
- Gene Aliases
- Chromosome Location
- Chr. 10: 66226015 - 66229293 on Build Rnor_6.0
- UniGene ID
- Rn.64588
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_012976.2 |
NP_037108.1
|
2, 3 |
163 |
|
GenBank mRNA |
FQ225275.1 |
- |
2, 3 |
157 |
|
FQ225984.1 |
- |
2, 3 |
179 |
|
FQ226981.1 |
- |
2, 3 |
184 |
|
FQ227276.1 |
- |
2, 3 |
178 |
|
FQ228081.1 |
- |
2, 3 |
183 |
|
FQ230182.1 |
- |
2, 3 |
183 |
|
FQ231538.1 |
- |
2, 3 |
182 |
|
FQ231564.1 |
- |
2, 3 |
193 |
|
FQ232247.1 |
- |
2, 3 |
204 |
|
FQ232513.1 |
- |
2, 3 |
186 |
|
FQ233614.1 |
- |
2, 3 |
204 |
|
FQ234601.1 |
- |
2, 3 |
186 |
|
FQ234845.1 |
- |
2, 3 |
206 |
|
L21711.1 |
- |
2, 3 |
172 |
|
L36862.1 |
- |
2, 3 |
163 |
|
View Full Product Details