Gene Transcripts

Gene Symbol
Lgals5
Entrez Gene ID
25475
Gene Name
lectin, galactose binding, soluble 5
Gene Aliases
Chromosome Location
Chr. 10: 66226015 - 66229293 on Build Rnor_6.0
UniGene ID
Rn.64588
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012976.2 NP_037108.1 4 378
GenBank mRNA FQ225275.1 - 4 372
FQ225984.1 - 4 394
FQ226981.1 - 4 399
FQ228081.1 - 4 398
FQ230182.1 - 4 398
FQ231538.1 - 4 397
FQ231564.1 - 4 408
FQ232247.1 - 4 419
FQ232513.1 - 4 401
FQ233614.1 - 4 419
FQ234601.1 - 4 401
FQ234845.1 - 4 421
L21711.1 - 4 387
L36862.1 - 4 378

View Full Product Details