Gene Transcripts
- Gene Symbol
- TROVE2
- Entrez Gene ID
- 6738
- Gene Name
- TROVE domain family member 2
- Gene Aliases
- RO60, RORNP, SSA2
- Chromosome Location
- Chr. 1: 193059422 - 193091777 on Build GRCh38
- UniGene ID
- Hs.288178
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001042369.2 |
NP_001035828.1
|
4, 5 |
1326 |
|
NM_001042370.2 |
NP_001035829.2
|
4, 5 |
1326 |
|
NM_001173524.1 |
NP_001166995.1
|
4, 5 |
1332 |
|
NM_001173525.1 |
NP_001166996.1
|
4, 5 |
1326 |
|
NM_004600.5 |
NP_004591.2
|
4, 5 |
1326 |
|
NR_033393.1 |
|
3, 4 |
731 |
|
XM_006711495.3 |
XP_006711558.1
|
5, 6 |
1398 |
|
XM_006711496.3 |
XP_006711559.1
|
6, 7 |
1574 |
|
XM_006711497.3 |
XP_006711560.1
|
5, 6 |
1508 |
|
XM_011509922.2 |
XP_011508224.1
|
4, 5 |
907 |
|
XM_017002180.1 |
XP_016857669.1
|
5, 6 |
1207 |
|
XM_017002181.1 |
XP_016857670.1
|
5, 6 |
1436 |
|
XM_017002182.1 |
XP_016857671.1
|
4, 5 |
1141 |
|
XM_017002183.1 |
XP_016857672.1
|
2, 3 |
478 |
|
GenBank mRNA |
AK314594.1 |
- |
4, 5 |
1119 |
|
AY205314.1 |
- |
4, 5 |
1140 |
|
AY205315.1 |
- |
4, 5 |
1140 |
|
BC036658.2 |
- |
4, 5 |
1168 |
4821776 |
BX394323.2 |
- |
3, 4 |
304 |
|
CR936683.1 |
- |
3, 4 |
700 |
|
J04137.1 |
- |
3, 4 |
995 |
|
M25077.1 |
- |
4, 5 |
1140 |
|
View Full Product Details