Gene Transcripts

Gene Symbol
SULT2A1
Entrez Gene ID
6822
Gene Name
sulfotransferase family 2A member 1
Gene Aliases
DHEA-ST, DHEAS, HST, ST2, ST2A1, ST2A3, STD, hSTa
Chromosome Location
Chr. 19: 47870466 - 47886397 on Build GRCh38
UniGene ID
Hs.515835
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003167.3 NP_003158.2 3 549
GenBank mRNA AK289380.1 - 3 491
AK313415.1 - 3 476
BC020755.1 - 3 466 4733628
BG563083.1 - 3 529
BX509490.1 - 3 550
L02337.1 - 3 474
L20000.1 - 3 461
S43859.1 - 3 474
S43861.1 - 2 243
U08024.1 - 3 429
U08025.1 - 3 473
X70222.1 - 3 461
X84816.1 - 3 454

View Full Product Details