Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_020783.3 | NP_065834.1 | 2 | 944 | |
GenBank mRNA | AB037763.1 | - | 2 | 761 | |
AF299075.1 | - | 2 | 668 | ||
AK291154.1 | - | 2 | 784 | ||
AK293791.1 | - | 2 | 730 | ||
BC036538.2 | - | 2 | 673 | 5260337 |