Gene Transcripts

Gene Symbol
Rtn4r
Entrez Gene ID
113912
Gene Name
reticulon 4 receptor
Gene Aliases
NgR, NgR1, Nogor
Chromosome Location
Chr. 11: 86992389 - 87017112 on Build Rnor_6.0
UniGene ID
Rn.229952
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053613.1 NP_446065.1 2 240
XM_008768839.1 XP_008767061.1 2 773
GenBank mRNA AF462390.1 - 2 240
AY028438.1 - 2 240

View Full Product Details