Gene Transcripts
- Gene Symbol
- Pou2f1
- Entrez Gene ID
- 171068
- Gene Name
- POU class 2 homeobox 1
- Gene Aliases
- Chromosome Location
- Chr. 13: 84080022 - 84217356 on Build Rnor_6.0
- UniGene ID
- Rn.46306
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001100639.1 |
NP_001094109.1
|
6, 7 |
601 |
|
XM_008769591.1 |
XP_008767813.1
|
6, 7 |
2727 |
|
XM_008769592.1 |
XP_008767814.1
|
6, 7 |
862 |
|
XM_008769593.1 |
XP_008767815.1
|
8, 9 |
1061 |
|
XM_008769594.1 |
XP_008767816.1
|
7, 8 |
963 |
|
XM_008769595.1 |
XP_008767817.1
|
8, 9 |
996 |
|
XM_008769596.1 |
XP_008767818.1
|
8, 9 |
3171 |
|
XM_008769597.1 |
XP_008767819.1
|
7, 8 |
3073 |
|
XM_008769598.1 |
XP_008767820.1
|
6, 7 |
2972 |
|
XM_008769599.1 |
XP_008767821.1
|
6, 7 |
625 |
|
GenBank mRNA |
DV214198.1 |
- |
6, 7 |
601 |
|
U17013.1 |
- |
1, 2 |
191 |
|
View Full Product Details