Gene Transcripts

Gene Symbol
Cst12
Entrez Gene ID
266776
Gene Name
cystatin 12
Gene Aliases
Chromosome Location
Chr. 3: 143119697 - 143123557 on Build Rnor_6.0
UniGene ID
Rn.58124
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153734.1 NP_714956.1 1 108
GenBank mRNA AF440736.1 - 1 108

View Full Product Details