Gene Transcripts
- Gene Symbol
- Cyp4f4
- Entrez Gene ID
- 286904
- Gene Name
- cytochrome P450, family 4, subfamily f, polypeptide 4
- Gene Aliases
- Chromosome Location
- Chr. 7: 14891062 - 14907250 on Build Rnor_6.0
- UniGene ID
- Rn.10170
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_173123.1 |
NP_775146.1
|
7 |
884 |
|
GenBank mRNA |
U39206.1 |
- |
7 |
884 |
|
View Full Product Details