Gene Transcripts

Gene Symbol
Hid1
Entrez Gene ID
287822
Gene Name
HID1 domain containing
Gene Aliases
RGD1311422
Chromosome Location
Chr. 10: 103899168 - 103919646 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006247774.2 XP_006247836.1 9 1270
XM_213523.8 XP_213523.6 9 1266

View Full Product Details