Gene Transcripts

Gene Symbol
Lgals12
Entrez Gene ID
293710
Gene Name
lectin, galactoside-binding, soluble, 12
Gene Aliases
Chromosome Location
Chr. 1: 222891626 - 222903183 on Build Rnor_6.0
UniGene ID
Rn.28915
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106333.1 NP_001099803.1 1, 2 534
XM_006230763.2 XP_006230825.1 2, 3 1101
XM_006230764.2 XP_006230826.1 2, 3 364
XM_008760110.1 XP_008758332.1 2, 3 1313
XM_008760111.1 XP_008758333.1 2, 3 1297
XM_008760112.1 XP_008758334.1 1, 2 463

View Full Product Details