Gene Transcripts

Gene Symbol
VLDLR
Entrez Gene ID
7436
Gene Name
very low density lipoprotein receptor
Gene Aliases
CAMRQ1, CARMQ1, CHRMQ1, VLDL-R, VLDLRCH
Chromosome Location
Chr. 9: 2621679 - 2656103 on Build GRCh38
UniGene ID
Hs.370422
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001018056.2 NP_001018066.1 7, 8 1484
NM_001322225.1 NP_001309154.1 6, 7 1361
NM_001322226.1 NP_001309155.1 6, 7 1361
NM_003383.4 NP_003374.3 7, 8 1484
GenBank mRNA AB208822.1 - 8, 9 1240
AK092381.1 - 5, 6 751
AK095744.1 - 3, 4 564
BC136562.1 - 7, 8 1205 9020552
BC142653.1 - 7, 8 1146 40148037
BC144245.1 - 7, 8 1205 9052765
D16493.1 - 7, 8 1138
D16494.1 - 7, 8 1069
L20470.1 - 7, 8 1459
L22431.1 - 7, 8 1181
S73849.1 - 7, 8 1153

View Full Product Details