Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_014146.3 | NP_054865.2 | 11 | 942 | |
NM_032463.2 | NP_115852.1 | 12 | 1061 | ||
NM_032464.2 | NP_115853.2 | 12 | 1131 | ||
XM_011516558.2 | XP_011514860.1 | 12 | 971 | ||
GenBank mRNA | AF161531.1 | - | 11 | 691 | |
AF252611.1 | - | 12 | 882 | ||
AF252612.1 | - | 10 | 637 | ||
AF252613.1 | - | 11 | 678 | ||
AF252614.1 | - | 12 | 812 | ||
AF257135.1 | - | 10 | 613 | ||
AK002099.1 | - | 12 | 921 | ||
AK092904.1 | - | 12 | 1272 | ||
AK290074.1 | - | 12 | 809 | ||
AK290916.1 | - | 11 | 690 | ||
AY190023.1 | - | 10 | 623 | ||
BC001609.1 | - | 12 | 943 | 3543310 | |
BC009204.2 | - | 10 | 615 | 4054137 |