Gene Transcripts

Gene Symbol
Sdr16c5
Entrez Gene ID
297805
Gene Name
short chain dehydrogenase/reductase family 16C, member 5
Gene Aliases
RGD1565999, Rdhe2
Chromosome Location
Chr. 5: 16904926 - 16930076 on Build Rnor_6.0
UniGene ID
Rn.17686
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106634.1 NP_001100104.1 1 273
XM_006237836.2 XP_006237898.1 2 160

View Full Product Details