Gene Transcripts

Gene Symbol
Lemd3
Entrez Gene ID
680066
Gene Name
LEM domain containing 3
Gene Aliases
RGD1561001
Chromosome Location
Chr. 7: 62977478 - 63045783 on Build Rnor_6.0
UniGene ID
Rn.22132
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191000.1 NP_001177929.1 1 1437
XM_006241410.2 XP_006241472.1 1 1492

View Full Product Details