Gene Transcripts

Gene Symbol
Troap
Entrez Gene ID
300219
Gene Name
trophinin associated protein
Gene Aliases
Chromosome Location
Chr. 7: 140758584 - 140766423 on Build Rnor_6.0
UniGene ID
Rn.18273
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271243.1 NP_001258172.1 2 166
XM_006257348.2 XP_006257410.1 2 197
GenBank mRNA FQ226442.1 - 2 185

View Full Product Details