Gene Transcripts

Gene Symbol
Tns1
Entrez Gene ID
301509
Gene Name
tensin 1
Gene Aliases
Tns
Chromosome Location
Chr. 9: 81191792 - 81401027 on Build Rnor_6.0
UniGene ID
Rn.228393
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191810.1 NP_001178739.1 19 2166
XM_008767220.1 XP_008765442.1 19 2206

View Full Product Details