Gene Transcripts

Gene Symbol
SHFM1
Entrez Gene ID
7979
Gene Name
split hand/foot malformation (ectrodactyly) type 1
Gene Aliases
DSS1, ECD, SEM1, SHFD1, SHSF1, Shfdg1
Chromosome Location
Chr. 7: 96688767 - 96709891 on Build GRCh38
UniGene ID
Hs.489201
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006304.1 NP_006295.1 3 393
GenBank mRNA AK094899.1 - 2 2667
AK291070.1 - 3 373
BC032782.1 - 3 353 5166837
U41515.1 - 3 393

View Full Product Details