Gene Transcripts

Gene Symbol
Slfn5
Entrez Gene ID
303377
Gene Name
schlafen family member 5
Gene Aliases
Chromosome Location
Chr. 10: 70298092 - 70312065 on Build Rnor_6.0
UniGene ID
Rn.199127
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008768150.1 XP_008766372.1 2 1423
XM_220775.6 XP_220775.3 3 1290

View Full Product Details