Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_006767.3 | NP_006758.2 | 16 | 1907 | |
GenBank mRNA | AK296347.1 | - | 15 | 1788 | |
AK313502.1 | - | 11 | 1309 | ||
BC026214.1 | - | 16 | 1809 | 4399716 | |
CT841521.1 | - | 16 | 1823 | ||
D38496.1 | - | 16 | 1802 |