Gene Transcripts

Gene Symbol
Pbrm1
Entrez Gene ID
306254
Gene Name
polybromo 1
Gene Aliases
RGD1565549
Chromosome Location
Chr. 16: 7103989 - 7202899 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006252685.2 XP_006252747.1 29 5203
XM_006252686.2 XP_006252748.1 28 4465
XM_006252696.2 XP_006252758.1 27 4341
XM_008771034.1 XP_008769256.1 28 5411
XM_008771035.1 XP_008769257.1 28 4813
XM_008771036.1 XP_008769258.1 28 5408
XM_008771037.1 XP_008769259.1 28 5408
XM_008771038.1 XP_008769260.1 28 5402
XM_008771039.1 XP_008769261.1 28 5399
XM_008771040.1 XP_008769262.1 27 5366
XM_008771041.1 XP_008769263.1 27 5363
XM_008771042.1 XP_008769264.1 28 5079
XM_008771043.1 XP_008769265.1 28 5076
XM_008771044.1 XP_008769266.1 27 5315
XM_008771045.1 XP_008769267.1 28 5411
XM_008771046.1 XP_008769268.1 28 5408
XM_008771047.1 XP_008769269.1 28 5411
XM_008771048.1 XP_008769270.1 27 5363
XM_008771049.1 XP_008769271.1 26 5267
XM_008771050.1 XP_008769272.1 28 5411
XM_008771051.1 XP_008769273.1 28 5408
XM_008771052.1 XP_008769274.1 27 5363

View Full Product Details