Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001013136.1 | NP_001013154.1 | 3 | 645 | |
XM_006252898.2 | XP_006252960.1 | 4 | 519 | ||
XM_006252899.2 | XP_006252961.1 | 4 | 556 | ||
GenBank mRNA | BC087111.1 | - | 3 | 645 |