Gene Transcripts

Gene Symbol
Otud4
Entrez Gene ID
307774
Gene Name
OTU deubiquitinase 4
Gene Aliases
RGD1305606
Chromosome Location
Chr. 19: 31895753 - 31942516 on Build Rnor_6.0
UniGene ID
Rn.228676
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191700.1 NP_001178629.1 2 431
XM_006255402.2 XP_006255464.1 2 767
XM_008772493.1 XP_008770715.1 2 943
XM_008772494.1 XP_008770716.1 2 499
XM_008772495.1 XP_008770717.1 3 547

View Full Product Details