Gene Transcripts

Gene Symbol
Shprh
Entrez Gene ID
308282
Gene Name
SNF2 histone linker PHD RING helicase, E3 ubiquitin protein ligase
Gene Aliases
Chromosome Location
Chr. 1: 5240139 - 5313064 on Build Rnor_6.0
UniGene ID
Rn.2515
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107470.1 NP_001100940.1 9 1877
XM_006227639.2 XP_006227701.1 9 1921
XM_008758609.1 XP_008756831.1 9 1921

View Full Product Details