Gene Transcripts
- Gene Symbol
- PIP5K1A
- Entrez Gene ID
- 8394
- Gene Name
- phosphatidylinositol-4-phosphate 5-kinase type 1 alpha
- Gene Aliases
- Chromosome Location
- Chr. 1: 151198003 - 151249536 on Build GRCh38
- UniGene ID
- Hs.655131
- Species
- Homo sapiens
- Gene Symbol
- PIP5K1P1
- Entrez Gene ID
- 206426
- Gene Name
- phosphatidylinositol-4-phosphate 5-kinase type 1 pseudogene 1
- Gene Aliases
- Chromosome Location
- Chr. 6: 7986102 - 7990344 on Build GRCh38
- UniGene ID
- Hs.676966
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027712.1 |
|
1 |
1063 |
|
GenBank mRNA |
AK312819.1 |
- |
1 |
668 |
|
BC028580.2 |
- |
1 |
1062 |
|
- Gene Symbol
- PIPSL
- Entrez Gene ID
- 266971
- Gene Name
- PIP5K1A and PSMD4-like, pseudogene
- Gene Aliases
- PIP5K1L1, PIP5K1P3, PSMD4P2, bA429H9.1
- Chromosome Location
- Chr. 10: 93958140 - 93961915 on Build GRCh38
- UniGene ID
- Hs.149255
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002319.2 |
|
1 |
1027 |
|
View Full Product Details