Gene Transcripts

Gene Symbol
Ppapdc1a
Entrez Gene ID
309014
Gene Name
phosphatidic acid phosphatase type 2 domain containing 1A
Gene Aliases
RGD1306289
Chromosome Location
Chr. 1: 202432366 - 202560628 on Build Rnor_6.0
UniGene ID
Rn.53410
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191631.1 NP_001178560.1 6, 7 688

View Full Product Details