Gene Transcripts

Gene Symbol
Rbm46
Entrez Gene ID
310548
Gene Name
RNA binding motif protein 46
Gene Aliases
RGD1560205
Chromosome Location
Chr. 2: 181828932 - 181874223 on Build Rnor_6.0
UniGene ID
Rn.145040
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135717.1 NP_001129189.1 3 569
XM_006232530.2 XP_006232592.1 3 564

View Full Product Details