Gene Transcripts

Gene Symbol
Cc2d1b
Entrez Gene ID
313478
Gene Name
coiled-coil and C2 domain containing 1B
Gene Aliases
RGD1306630
Chromosome Location
Chr. 5: 128215711 - 128229762 on Build Rnor_6.0
UniGene ID
Rn.3265
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001270984.1 NP_001257913.1 15 1828
GenBank mRNA FQ212419.1 - 6 2549
FQ222909.1 - 12 1430

View Full Product Details