Gene Transcripts

Gene Symbol
Nudt7
Entrez Gene ID
361413
Gene Name
nudix (nucleoside diphosphate linked moiety X)-type motif 7
Gene Aliases
Chromosome Location
Chr. 19: 46455766 - 46470074 on Build Rnor_6.0
UniGene ID
Rn.99447
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108450.1 NP_001101920.1 3 1933

Gene Symbol
LOC102555567
Entrez Gene ID
102555567
Gene Name
peroxisomal coenzyme A diphosphatase NUDT7-like
Gene Aliases
Chromosome Location
Chr. 19: 46455685 - 46470070 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006255677.2 XP_006255739.2 2 249

View Full Product Details