Gene Transcripts

Gene Symbol
Pld4
Entrez Gene ID
362792
Gene Name
phospholipase D family, member 4
Gene Aliases
Chromosome Location
Chr. 6: 137323713 - 137331231 on Build Rnor_6.0
UniGene ID
Rn.6379
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001126288.1 NP_001119760.1 2 153
GenBank mRNA BC161906.1 - 2 61

View Full Product Details