Gene Transcripts

Gene Symbol
Mum1
Entrez Gene ID
362838
Gene Name
melanoma associated antigen (mutated) 1
Gene Aliases
Chromosome Location
Chr. 7: 12331288 - 12348220 on Build Rnor_6.0
UniGene ID
Rn.92148
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108736.2 NP_001102206.2 10 1510
XM_006240973.2 XP_006241035.1 10 1546
XM_006240974.2 XP_006241036.1 10 1579
XM_006240975.2 XP_006241037.1 10 2500
GenBank mRNA BC160828.1 - 10 1513
CB784307.1 - 5 377

View Full Product Details