Gene Transcripts

Gene Symbol
BTAF1
Entrez Gene ID
9044
Gene Name
B-TFIID TATA-box binding protein associated factor 1
Gene Aliases
MOT1, TAF(II)170, TAF172, TAFII170
Chromosome Location
Chr. 10: 91923769 - 92030998 on Build GRCh38
UniGene ID
Hs.500526
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003972.2 NP_003963.1 33, 34 4817
XM_011540326.1 XP_011538628.1 33, 34 4916
XM_011540327.2 XP_011538629.1 30, 31 4582
XM_011540328.2 XP_011538630.1 30, 31 4410
XM_017016877.1 XP_016872366.1 32, 33 4879
XM_017016878.1 XP_016872367.1 32, 33 4709
XM_017016879.1 XP_016872368.1 32, 33 4833
XM_017016880.1 XP_016872369.1 32, 33 4837
XM_017016881.1 XP_016872370.1 31, 32 4686
GenBank mRNA AB208843.1 - 21, 22 3211
AF038362.1 - 33, 34 4836
AJ001017.2 - 33, 34 4731
AK303554.1 - 11, 12 1494
BC029930.1 - 9, 10 1212
BC112201.1 - 33, 34 4801 8327669

View Full Product Details