Gene Transcripts

Gene Symbol
ATP1B3
Entrez Gene ID
483
Gene Name
ATPase Na+/K+ transporting subunit beta 3
Gene Aliases
ATPB-3, CD298
Chromosome Location
Chr. 3: 141876628 - 141926540 on Build GRCh38
UniGene ID
Hs.477789
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK131061.1 - 2 321
AK293697.1 - 3 416
BC011835.2 - 2 313 3944210
BI462090.1 - 2 353
HY040306.1 - 2 372
U51478.1 - 2 190

View Full Product Details