Gene Transcripts

Gene Symbol
Epb41l4b
Entrez Gene ID
500464
Gene Name
erythrocyte membrane protein band 4.1 like 4B
Gene Aliases
RGD1562988
Chromosome Location
Chr. 5: 74105491 - 74191167 on Build Rnor_6.0
UniGene ID
Rn.21106
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001127565.1 NP_001121037.1 12 1218
GenBank mRNA BC166749.1 - 12 1218

View Full Product Details