Gene Transcripts

Gene Symbol
Lsm11
Entrez Gene ID
501688
Gene Name
LSM11, U7 small nuclear RNA associated
Gene Aliases
RGD1565523
Chromosome Location
Chr. 10: 31023561 - 31041741 on Build Rnor_6.0
UniGene ID
Rn.106829
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001173384.1 NP_001166855.1 4 1049
XM_006246151.2 XP_006246213.1 3 1080
GenBank mRNA BC158836.1 - 4 1049
FQ223124.1 - 4 1085

View Full Product Details