Gene Transcripts

Gene Symbol
Vom2r60
Entrez Gene ID
501794
Gene Name
vomeronasal 2 receptor, 60
Gene Aliases
RGD1560561
Chromosome Location
Chr. 12: 4054395 - 4436352 on Build Rnor_6.0
UniGene ID
Rn.217624
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099480.1 NP_001092950.1 3 357

View Full Product Details