Gene Transcripts

Gene Symbol
Vom2r49
Entrez Gene ID
688392
Gene Name
vomeronasal 2 receptor, 49
Gene Aliases
Chromosome Location
Chr. 6: 56732891 - 56775529 on Build Rnor_6.0
UniGene ID
Rn.217630
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099515.1 NP_001092985.1 3 935

Gene Symbol
Vom2r50
Entrez Gene ID
500312
Gene Name
vomeronasal 2 receptor, 50
Gene Aliases
Chromosome Location
Chr. 4: 156482446 - 156504818 on Build Rnor_6.0
UniGene ID
Rn.134207
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099507.1 NP_001092977.1 3 944

Gene Symbol
Vom2r51
Entrez Gene ID
502891
Gene Name
vomeronasal 2 receptor, 51
Gene Aliases
RGD1561712
Chromosome Location
Chr. 4: 156515299 - 156563438 on Build Rnor_6.0
UniGene ID
Rn.217944
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099510.1 NP_001092980.1 3 935

View Full Product Details