Gene Transcripts

Gene Symbol
Syce3
Entrez Gene ID
689933
Gene Name
synaptonemal complex central element protein 3
Gene Aliases
Chromosome Location
Chr. 7: 130360511 - 130386668 on Build Rnor_6.0
UniGene ID
Rn.33202
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135253.1 NP_001128725.1 2 421
XM_006242244.2 XP_006242306.1 3 696
XM_006242246.2 XP_006242308.1 3 571

View Full Product Details