Gene Transcripts

Gene Symbol
Nudt13
Entrez Gene ID
682978
Gene Name
nudix (nucleoside diphosphate linked moiety X)-type motif 13
Gene Aliases
Chromosome Location
Chr. 15: 4434341 - 4454878 on Build Rnor_6.0
UniGene ID
Rn.160892
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001127636.1 NP_001121108.1 2, 3 168
GenBank mRNA BC166528.1 - 2, 3 168

View Full Product Details