Gene Transcripts

Gene Symbol
Stra13
Entrez Gene ID
688314
Gene Name
stimulated by retinoic acid 13
Gene Aliases
Chromosome Location
Chr. 10: 109890904 - 109893894 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003750986.3 XP_003751034.2 4 335
XM_008768490.1 XP_008766712.1 3 434
XM_008768491.1 XP_008766713.1 4 512

View Full Product Details