Gene Transcripts

Gene Symbol
ZBTB39
Entrez Gene ID
9880
Gene Name
zinc finger and BTB domain containing 39
Gene Aliases
ZNF922
Chromosome Location
Chr. 12: 56998833 - 57006513 on Build GRCh38
UniGene ID
Hs.591025
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014830.2 NP_055645.1 2 2176
GenBank mRNA AB002350.1 - 2 2109
AK292468.1 - 2 2176
BC140890.1 - 1 2079 9021758
BC146867.1 - 1 2079 9056795
BC152461.1 - 2 2109 8862597

Gene Symbol
RDH16
Entrez Gene ID
8608
Gene Name
retinol dehydrogenase 16 (all-trans)
Gene Aliases
RODH-4, SDR9C8
Chromosome Location
Chr. 12: 56951431 - 56959374 on Build GRCh38
UniGene ID
Hs.134958
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK311025.1 - 2 504

View Full Product Details