Gene Transcripts

Gene Symbol
TELO2
Entrez Gene ID
9894
Gene Name
telomere maintenance 2
Gene Aliases
CLK2, TEL2, YHFS
Chromosome Location
Chr. 16: 1493344 - 1510459 on Build GRCh38
UniGene ID
Hs.271044
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016111.3 NP_057195.2 19, 20 2575
XM_011522773.2 XP_011521075.1 20, 21 2752
XM_011522774.2 XP_011521076.1 20, 21 2950
XM_011522775.2 XP_011521077.1 20, 21 2733
XM_011522776.2 XP_011521078.1 20, 21 2820
XM_017023914.1 XP_016879403.1 19, 20 2547
GenBank mRNA AB014583.3 - 19, 20 2563
AK001786.1 - 9, 10 1011
AL080126.1 - 19, 20 2506
AL137394.1 - 19, 20 2506
BC017188.2 - 18, 19 2331 3139743
BC028332.1 - 2, 3 100

View Full Product Details