Gene Transcripts

Gene Symbol
Rnpc3
Entrez Gene ID
691538
Gene Name
RNA-binding region (RNP1, RRM) containing 3
Gene Aliases
Chromosome Location
Chr. 2: 216481457 - 216510054 on Build Rnor_6.0
UniGene ID
Rn.17587
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100810.1 NP_001094280.1 4, 5 496
XM_006233211.2 XP_006233273.1 4, 5 874
XM_008761424.1 XP_008759646.1 4, 5 874
XM_008761425.1 XP_008759647.1 4, 5 875
GenBank mRNA BC098742.1 - 2, 3 174
BC166571.1 - 4, 5 496 7320328

View Full Product Details