Gene Transcripts

Gene Symbol
Gpr17
Entrez Gene ID
767613
Gene Name
G protein-coupled receptor 17
Gene Aliases
Chromosome Location
Chr. 18: 24733306 - 24738283 on Build Rnor_6.0
UniGene ID
Rn.212749
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001071777.1 NP_001065245.1 1 775
XM_006254604.2 XP_006254666.1 2 2292
GenBank mRNA DQ777767.1 - 1 775

View Full Product Details