Gene Transcripts

Gene Symbol
NFKBIB
Entrez Gene ID
4793
Gene Name
NFKB inhibitor beta
Gene Aliases
IKBB, TRIP9
Chromosome Location
Chr. 19: 38899503 - 38908894 on Build GRCh38
UniGene ID
Hs.9731
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001243116.1 NP_001230045.1 6 938
NM_002503.4 NP_002494.2 6 1180
NR_040515.1 5 2195
XM_006723226.2 XP_006723289.2 6 1116
XM_006723227.3 XP_006723290.3 6 1079
GenBank mRNA AK057862.1 - 5 2146
BC015528.1 - 6 1111 3903794
BX111397.1 - 1 321

View Full Product Details