Gene Transcripts

Gene Symbol
RPS27
Entrez Gene ID
6232
Gene Name
ribosomal protein S27
Gene Aliases
MPS-1, MPS1, S27
Chromosome Location
Chr. 1: 153990763 - 153992155 on Build GRCh38
UniGene ID
Hs.546291
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001030.4 NP_001021.1 1 281
GenBank mRNA AV763942.1 - 1 282
BC002658.2 - 1 264 3607957
BC070219.1 - 282 6744779
CN304370.1 - 1 277
L19739.1 - 4 267
U57847.1 - 4 260

Gene Symbol
ZNF320
Entrez Gene ID
162967
Gene Name
zinc finger protein 320
Gene Aliases
ZFPL
Chromosome Location
Chr. 19: 52863790 - 52897695 on Build GRCh38
UniGene ID
Hs.369632
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BC064408.1 - 1421

View Full Product Details