Gene Transcripts

Gene Symbol
FRAT1
Entrez Gene ID
10023
Gene Name
frequently rearranged in advanced T-cell lymphomas 1
Gene Aliases
Chromosome Location
Chr. 10: 97319265 - 97321915 on Build GRCh38
UniGene ID
Hs.126057
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005479.3 NP_005470.2 1 1487
GenBank mRNA AB074890.1 - 1 1410
U58975.2 - 1 1486

View Full Product Details